Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009367.3 | NP_033393.2 | 4 | 1911 | ||
GenBank mRNA | AK029306.1 | - | 2 | 123 | ||
AK159690.1 | - | 4 | 475 | |||
BC011055.1 | - | 2 | 274 | 3585774 | ||
BC011170.1 | - | 4 | 819 | 4035366 | ||
X57413.1 | - | 4 | 1910 |