Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp322a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111107.2 | NP_001104577.1 | 5 | 1392 | ||
NM_001286155.1 | NP_001273084.1 | 5 | 1670 | |||
NM_001286156.1 | NP_001273085.1 | 4 | 1595 | |||
NM_172586.4 | NP_766174.1 | 4 | 1317 | |||
GenBank mRNA | AK036302.1 | - | 4 | 1318 | ||
AK036877.1 | - | 4 | 1318 | |||
AK162608.1 | - | 4 | 1295 | |||
AK170259.1 | - | 4 | 1308 | |||
AK170525.1 | - | 4 | 1312 | |||
BC017143.1 | - | 1 | 881 | |||
BC032268.1 | - | 1 | 951 | |||
BC043711.1 | - | 4 | 1279 | 5369339 |