Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011578.3 | NP_035708.2 | 7 | 1279 | ||
GenBank mRNA | AF039601.1 | - | 7 | 1079 | ||
AK133234.1 | - | 5 | 789 | |||
AK166082.1 | - | 7 | 1280 | |||
BC070428.1 | - | 7 | 1204 | 6410554 |