Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011578.3 | NP_035708.2 | 8 | 1372 | ||
GenBank mRNA | AF039601.1 | - | 8 | 1172 | ||
AK133234.1 | - | 6 | 882 | |||
BC070428.1 | - | 8 | 1297 | 6410554 |