Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122989.1 | NP_001116461.1 | 6, 7 | 549 | ||
NM_172587.3 | NP_766175.3 | 6, 7 | 813 | |||
GenBank mRNA | AK041155.1 | - | 6, 7 | 535 | ||
AK044332.1 | - | 6, 7 | 1115 | |||
BC049794.1 | - | 6, 7 | 755 | 6514098 | ||
BC057357.1 | - | 6, 7 | 704 | 6835123 | ||
CN720173.1 | - | 6, 7 | 467 |