Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122989.1 | NP_001116461.1 | 4 | 352 | ||
NM_172587.3 | NP_766175.3 | 4 | 616 | |||
GenBank mRNA | AK041155.1 | - | 4 | 338 | ||
AK044332.1 | - | 4 | 918 | |||
AK161345.1 | - | 4 | 617 | |||
BC049794.1 | - | 4 | 558 | 6514098 | ||
BC057357.1 | - | 4 | 507 | 6835123 | ||
CN720173.1 | - | 4 | 270 |