Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164078.1 | NP_001157550.1 | 6 | 684 | ||
NM_001164079.1 | NP_001157551.1 | 6 | 678 | |||
NM_011585.4 | NP_035715.1 | 7 | 711 | |||
GenBank mRNA | AK028340.1 | - | 5 | 351 | ||
AK033792.1 | - | 1 | 2399 | |||
AK088486.1 | - | 7 | 647 | |||
AK144222.1 | - | 7 | 664 | |||
AK219644.1 | - | 4 | 196 | |||
BC023813.1 | - | 6 | 593 | 5353938 | ||
BC027826.1 | - | 5 | 2648 | |||
BC046812.1 | - | 6 | 567 | 5702183 | ||
U00689.1 | - | 7 | 652 | |||
U55862.1 | - | 7 | 519 |