Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173392.4 | NP_775568.1 | 3 | 1001 | ||
GenBank mRNA | AK045090.1 | - | 3 | 1059 | ||
AK122241.1 | - | 3 | 982 | |||
AK170435.1 | - | 3 | 1020 | |||
BC042669.1 | - | 3 | 999 | 4235024 |