Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145886.1 | NP_001139358.1 | 16, 17 | 3469 | ||
NM_001145887.1 | NP_001139359.1 | 1, 2 | 402 | |||
NM_009384.3 | NP_033410.2 | 16, 17 | 3504 | |||
GenBank mRNA | AK136856.1 | - | 1, 2 | 402 | ||
AK147392.1 | - | 16, 17 | 3470 | |||
AK161477.1 | - | 7, 8 | 811 | |||
AK162945.1 | - | 3, 4 | 458 | |||
U05245.1 | - | 16, 17 | 3507 |