Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272064.1 | NP_001258993.1 | 17, 18 | 2989 | ||
NM_001272076.1 | NP_001259005.1 | 17, 18 | 2671 | |||
NM_145076.4 | NP_659542.3 | 17, 18 | 3091 | |||
GenBank mRNA | AB221637.1 | - | 17, 18 | 2791 | ||
AK133144.1 | - | 17, 18 | 2671 | |||
AK166538.1 | - | 13, 14 | 2048 | |||
BC025482.1 | - | 8, 9 | 1186 | |||
BC056959.1 | - | 17, 18 | 3069 | 5698079 | ||
S78219.1 | - | 17, 18 | 2989 | |||
S78221.1 | - | 17, 18 | 3091 |