Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 9, 10 | 1174 | ||
GenBank mRNA | AK031041.1 | - | 9, 10 | 1177 | ||
AK142282.1 | - | 9, 10 | 860 | |||
AK146566.1 | - | 9, 10 | 908 | |||
AK164135.1 | - | 9, 10 | 920 | |||
BC019933.1 | - | 10, 11 | 910 | |||
BC047122.1 | - | 10, 11 | 910 | |||
BC052456.1 | - | 9, 10 | 871 | 6830607 | ||
BC053718.1 | - | 9, 10 | 890 |