Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 10, 11 | 1266 | ||
GenBank mRNA | AK031041.1 | - | 10, 11 | 1269 | ||
AK142282.1 | - | 10, 11 | 952 | |||
AK146566.1 | - | 10, 11 | 1000 | |||
AK164135.1 | - | 10, 11 | 1012 | |||
BC052456.1 | - | 10, 11 | 963 | 6830607 | ||
BC053718.1 | - | 10, 11 | 982 |