Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139299.2 | NP_647460.2 | 17 | 2146 | ||
GenBank mRNA | AB083111.1 | - | 17 | 2145 | ||
AK030512.1 | - | 6 | 715 |