Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172595.4 | NP_766183.1 | 1 | 170 | ||
GenBank mRNA | AK039965.1 | - | 1 | 134 | ||
AK048736.1 | - | 1 | 53 | |||
AK050979.1 | - | 1 | 85 | |||
AK052164.1 | - | 1 | 119 | |||
AK053953.1 | - | 1 | 86 | |||
BC120664.1 | - | 1 | 56 | 40129587 | ||
BC120666.1 | - | 1 | 56 | 40129589 | ||
BC144993.1 | - | 1 | 56 | 9053541 | ||
CJ063085.1 | - | 1 | 170 | |||
CJ066332.1 | - | 1 | 154 |