Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079849.2 | NP_001073318.1 | 4, 5 | 520 | ||
NM_145457.4 | NP_663432.1 | 4, 5 | 738 | |||
GenBank mRNA | AF128835.1 | - | 3, 4 | 420 | ||
AK040012.1 | - | 4, 5 | 521 | |||
BB202059.2 | - | 1, 2 | 109 | |||
BC019726.1 | - | 4, 5 | 709 | 4010170 | ||
BC051042.1 | - | 4, 5 | 695 | 4008643 | ||
CB840007.1 | - | 3, 4 | 493 | |||
CD363907.1 | - | 4, 5 | 496 |