Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033270.2 | NP_001028442.2 | 17 | 2489 | ||
GenBank mRNA | AK049322.1 | - | 8 | 1151 | ||
AK088400.1 | - | 17 | 2411 | |||
AK152233.1 | - | 17 | 2490 | |||
AK153567.1 | - | 17 | 2490 | |||
BC038373.1 | - | 18 | 2382 | |||
GU386352.1 | - | 17 | 2396 | |||
GU386353.2 | - | 17 | 2435 | |||
HM624050.1 | - | 17 | 2299 | |||
HM624051.1 | - | 18 | 2629 | |||
JQ073566.1 | - | 17 | 2281 | |||
JQ344322.1 | - | 17 | 2260 | |||
JQ347261.1 | - | 18 | 2359 | |||
JQ349031.1 | - | 18 | 2495 | |||
JQ349032.1 | - | 18 | 2456 | |||
JQ349033.1 | - | 18 | 2320 | |||
JX254908.1 | - | 18 | 2804 | |||
JX254909.1 | - | 18 | 2765 | |||
JX254910.1 | - | 18 | 2804 | |||
JX254911.1 | - | 18 | 2765 | |||
JX254912.1 | - | 18 | 2668 | |||
JX310364.1 | - | 18 | 2765 | |||
KF279520.1 | - | 19 | 2399 | |||
KF279521.1 | - | 19 | 2835 |