Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033270.2 | NP_001028442.2 | 21 | 3029 | ||
GenBank mRNA | AK049322.1 | - | 12 | 1691 | ||
AK088400.1 | - | 20 | 2699 | |||
BC038373.1 | - | 22 | 2922 | |||
GU386352.1 | - | 21 | 2936 | |||
GU386353.2 | - | 21 | 2975 | |||
HM624050.1 | - | 21 | 2839 | |||
HM624051.1 | - | 22 | 3169 | |||
JQ073566.1 | - | 21 | 2821 | |||
JQ344322.1 | - | 21 | 2800 | |||
JQ347261.1 | - | 22 | 2899 | |||
JQ349031.1 | - | 22 | 3035 | |||
JQ349032.1 | - | 22 | 2996 | |||
JQ349033.1 | - | 22 | 2860 | |||
JX254908.1 | - | 22 | 3344 | |||
JX254909.1 | - | 22 | 3305 | |||
JX254910.1 | - | 22 | 3344 | |||
JX254911.1 | - | 22 | 3305 | |||
JX254912.1 | - | 22 | 3208 | |||
JX310364.1 | - | 22 | 3305 | |||
KF279520.1 | - | 23 | 2939 | |||
KF279521.1 | - | 23 | 3375 |