Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289760.1 | NP_001276689.1 | 7 | 1350 | ||
NM_001289761.1 | NP_001276690.1 | 6 | 1269 | |||
NM_001289762.1 | NP_001276691.1 | 5 | 879 | |||
NM_011243.2 | NP_035373.1 | 5 | 1233 | |||
GenBank mRNA | AK043726.1 | - | 5 | 1587 | ||
AK078471.1 | - | 5 | 945 | |||
BC076597.1 | - | 5 | 1229 | 30608242 | ||
S56660.1 | - | 5 | 1129 |