Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009388.6 | NP_033414.1 | 8 | 1162 | ||
GenBank mRNA | AF195533.1 | - | 7 | 828 | ||
AK002627.1 | - | 8 | 1115 | |||
AK012794.1 | - | 8 | 1117 | |||
AK030446.1 | - | 8 | 1145 | |||
AK140965.1 | - | 8 | 1146 | |||
AK144146.1 | - | 8 | 1117 | |||
AK145677.1 | - | 8 | 1115 | |||
AK146157.1 | - | 8 | 1115 | |||
AK150139.1 | - | 8 | 1115 | |||
AK150769.1 | - | 8 | 1111 | |||
AK150844.1 | - | 8 | 1111 | |||
AK150856.1 | - | 8 | 1111 | |||
AK150901.1 | - | 8 | 1115 | |||
AK151916.1 | - | 8 | 1110 | |||
AK151969.1 | - | 8 | 1110 | |||
AK152245.1 | - | 8 | 1110 | |||
AK152460.1 | - | 8 | 1131 | |||
AK152886.1 | - | 8 | 1115 | |||
AK159922.1 | - | 8 | 1115 | |||
AK166763.1 | - | 8 | 1149 | |||
AK167084.1 | - | 8 | 1149 | |||
BC055336.1 | - | 8 | 1089 | 6413263 | ||
U05809.1 | - | 8 | 1110 |