Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081247.1 | NP_001074716.1 | 24, 25 | 3370 | ||
GenBank mRNA | BC053071.1 | - | 24, 25 | 3347 | ||
BC151002.1 | - | 24, 25 | 3280 | 9087915 | ||
BC151168.1 | - | 24, 25 | 3278 | 9088057 | ||
BC151169.1 | - | 24, 25 | 3280 | 9088058 |