Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144553.2 | NP_653136.2 | 19 | 2679 | ||
GenBank mRNA | AB076696.1 | - | 19 | 2679 | ||
AK085882.1 | - | 19 | 2460 | |||
AK135820.1 | - | 19 | 2486 | |||
BC027273.1 | - | 19 | 2400 | |||
BC043924.1 | - | 11 | 1305 | |||
BC058087.1 | - | 19 | 2413 | 6816660 |