Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276445.1 | NP_001263374.1 | 4 | 1044 | ||
NM_030682.2 | NP_109607.1 | 3 | 744 | |||
GenBank mRNA | AF316985.1 | - | 1 | 596 | ||
AK143721.1 | - | 4 | 1048 | |||
AK172870.1 | - | 1 | 372 | |||
AY009154.1 | - | 3 | 744 | |||
BC141319.1 | - | 3 | 806 | 9055931 | ||
BC141321.1 | - | 3 | 806 | 9055933 |