Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011604.3 | NP_035734.3 | 2 | 1000 | ||
GenBank mRNA | AB020808.1 | - | 1 | 888 | ||
AF314636.1 | - | 1 | 868 | |||
AK137502.1 | - | 2 | 955 | |||
AK154253.1 | - | 2 | 926 | |||
BC055366.1 | - | 2 | 1013 | 30056593 | ||
DQ790887.1 | - | 1 | 868 |