Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033271.4 | NP_001028443.1 | 3 | 481 | ||
GenBank mRNA | AK144251.1 | - | 3 | 481 | ||
AK152640.1 | - | 3 | 467 | |||
AK152658.1 | - | 3 | 467 | |||
AK159641.1 | - | 3 | 485 | |||
AK169026.1 | - | 2 | 906 | |||
BC027355.1 | - | 3 | 466 | |||
BC039161.1 | - | 3 | 424 | |||
BC049945.1 | - | 3 | 444 | |||
BC058171.1 | - | 3 | 430 | |||
BC106163.1 | - | 3 | 460 | 4913757 |