Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144843.4 | NP_659092.1 | 12 | 1567 | ||
GenBank mRNA | AB517623.1 | - | 13 | 1548 | ||
AK076218.1 | - | 12 | 1544 | |||
AK154859.1 | - | 12 | 1487 | |||
BC020019.1 | - | 12 | 1501 | 3596732 | ||
BC022979.1 | - | 2 | 608 |