Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177338.5 | NP_796312.2 | 3 | 365 | ||
GenBank mRNA | AK052729.1 | - | 3 | 375 | ||
AK089782.1 | - | 3 | 366 | |||
AK161592.1 | - | 3 | 365 | |||
BC051457.1 | - | 3 | 368 | 6398463 |