Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011609.4 | NP_035739.2 | 2 | 445 | ||
GenBank mRNA | AK154388.1 | - | 2 | 415 | ||
AK159989.1 | - | 2 | 369 | |||
AK211886.1 | - | 2 | 249 | |||
AY541589.1 | - | 2 | 126 | |||
AY541590.1 | - | 2 | 126 | |||
BC004599.1 | - | 2 | 361 | 3585060 | ||
BC052675.1 | - | 2 | 380 | 30061894 | ||
CK624043.1 | - | 2 | 445 | |||
L26349.1 | - | 2 | 413 | |||
M59377.1 | - | 2 | 239 | |||
M60468.1 | - | 2 | 338 | |||
X57796.1 | - | 2 | 416 | |||
X59238.1 | - | 2 | 338 |