Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011609.4 | NP_035739.2 | 3 | 570 | ||
GenBank mRNA | AK154388.1 | - | 3 | 540 | ||
AK159989.1 | - | 3 | 494 | |||
AK178992.1 | - | 1 | 41 | |||
AK206708.1 | - | 1 | 62 | |||
AK207044.1 | - | 1 | 41 | |||
AK211886.1 | - | 3 | 374 | |||
AY541589.1 | - | 3 | 251 | |||
AY541590.1 | - | 3 | 251 | |||
BC004599.1 | - | 3 | 486 | 3585060 | ||
BC052675.1 | - | 3 | 505 | 30061894 | ||
CK624043.1 | - | 3 | 570 | |||
L26349.1 | - | 3 | 538 | |||
M59377.1 | - | 3 | 364 | |||
M60468.1 | - | 3 | 463 | |||
X57796.1 | - | 3 | 541 | |||
X59238.1 | - | 3 | 463 |