Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022312.3 | NP_071707.2 | 23 | 4587 | ||
GenBank mRNA | AK039390.1 | - | 23 | 4588 | ||
AK078229.1 | - | 16 | 2265 | |||
BC132392.1 | - | 23 | 4221 | 40130669 | ||
BC138043.1 | - | 23 | 4221 | 8861062 |