Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011623.2 | NP_035753.2 | 19 | 2347 | ||
GenBank mRNA | AK033321.1 | - | 7 | 729 | ||
AK169723.1 | - | 4 | 359 | |||
D12513.1 | - | 19 | 2273 |