Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011625.1 | NP_035755.1 | 15, 16 | 3218 | ||
GenBank mRNA | AK183077.1 | - | 1, 2 | 109 | ||
BC034207.1 | - | 11, 12 | 2664 | |||
BC053092.1 | - | 15, 16 | 3244 | 5719383 | ||
BC053732.1 | - | 11, 12 | 2664 | |||
BC054788.1 | - | 15, 16 | 3218 | 6826065 | ||
U58881.1 | - | 2, 3 | 204 |