Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 10, 11 | 1398 | ||
GenBank mRNA | AK031918.1 | - | 6, 7 | 750 | ||
AK146885.1 | - | 10, 11 | 1464 | |||
AK150811.1 | - | 10, 11 | 1418 | |||
AK151413.1 | - | 10, 11 | 1415 | |||
AK153550.1 | - | 10, 11 | 1418 | |||
AK159974.1 | - | 10, 11 | 1423 | |||
AK160093.1 | - | 10, 11 | 1417 | |||
AK160827.1 | - | 838 | ||||
AK165975.1 | - | 10, 11 | 1418 | |||
AK168952.1 | - | 10, 11 | 1418 | |||
AK169090.1 | - | 10, 11 | 1418 | |||
AK183091.1 | - | 1, 2 | 67 | |||
AK209332.1 | - | 1, 2 | 67 | |||
AK212195.1 | - | 1, 2 | 67 | |||
BC010445.1 | - | 10, 11 | 1330 | 3587178 | ||
BC011439.1 | - | 10, 11 | 1404 | 3707817 | ||
J03297.1 | - | 10, 11 | 1398 | |||
M29652.1 | - | 2, 3 | 178 |