Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011634.3 | NP_035764.2 | 15 | 1502 | ||
GenBank mRNA | AK012786.1 | - | 15 | 1484 | ||
AK012948.1 | - | 15 | 1501 | |||
BC006929.1 | - | 14 | 3227 | |||
U77844.1 | - | 15 | 1507 |