Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133977.2 | NP_598738.1 | 16 | 2018 | ||
GenBank mRNA | AF440692.1 | - | 15 | 1948 | ||
AK085754.1 | - | 16 | 2003 | |||
AK142599.1 | - | 2008 | ||||
AK146549.1 | - | 16 | 2003 | |||
AK149559.1 | - | 16 | 2003 | |||
AK149595.1 | - | 16 | 2003 | |||
AK150782.1 | - | 16 | 2051 | |||
AK168368.1 | - | 16 | 2005 | |||
AK168405.1 | - | 16 | 2005 | |||
AK168419.1 | - | 16 | 2003 | |||
AK168938.1 | - | 16 | 2003 | |||
AK169187.1 | - | 16 | 2002 | |||
BC008559.1 | - | 14 | 1740 | |||
BC012313.1 | - | 16 | 2018 | 3981648 | ||
BC018573.1 | - | 11 | 1288 | |||
BC020295.1 | - | 16 | 1987 | |||
BC022986.1 | - | 16 | 1996 | 5123741 | ||
BC058216.1 | - | 16 | 1984 | |||
BC058218.1 | - | 16 | 1997 | |||
BC092046.1 | - | 16 | 1979 | 3594705 |