Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009439.1 | NP_033465.1 | 8 | 1352 | ||
GenBank mRNA | AK077252.1 | - | 8 | 1301 | ||
AK089933.1 | - | 8 | 1311 | |||
AK145467.1 | - | 8 | 1342 | |||
AK145488.1 | - | 8 | 1341 | |||
AK153505.1 | - | 8 | 1269 | |||
AK161632.1 | - | 3 | 236 | |||
AK164333.1 | - | 7 | 1578 | |||
AK164593.1 | - | 8 | 1341 | |||
AK165520.1 | - | 8 | 1287 | |||
AK166683.1 | - | 8 | 1312 | |||
AK166714.1 | - | 8 | 1340 | |||
AK167005.1 | - | 8 | 1327 | |||
AK167285.1 | - | 8 | 1307 | |||
AK169690.1 | - | 8 | 1341 | |||
BC003197.1 | - | 8 | 1352 | 3586427 |