Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009439.1 | NP_033465.1 | 2, 3 | 571 | ||
GenBank mRNA | AK031366.1 | - | 2, 3 | 561 | ||
AK077252.1 | - | 2, 3 | 520 | |||
AK089933.1 | - | 2, 3 | 529 | |||
AK145467.1 | - | 2, 3 | 561 | |||
AK145488.1 | - | 2, 3 | 560 | |||
AK153505.1 | - | 2, 3 | 488 | |||
AK164333.1 | - | 2, 3 | 535 | |||
AK164593.1 | - | 2, 3 | 560 | |||
AK165520.1 | - | 2, 3 | 506 | |||
AK166683.1 | - | 2, 3 | 531 | |||
AK166714.1 | - | 2, 3 | 559 | |||
AK167005.1 | - | 2, 3 | 546 | |||
AK167285.1 | - | 2, 3 | 526 | |||
AK169690.1 | - | 2, 3 | 560 | |||
AK178121.1 | - | 1, 2 | 89 | |||
BC003197.1 | - | 2, 3 | 571 | 3586427 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK003024.1 | - | 594 | Chr. - - [-] on Build |