Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009442.2 | NP_033468.2 | 5 | 1776 | ||
GenBank mRNA | AF237703.1 | - | 4 | 1728 | ||
AK140081.1 | - | 4 | 2994 | |||
BC059011.1 | - | 4 | 1743 | 6847867 | ||
X83974.1 | - | 6 | 1684 |