Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110265.1 | NP_001103735.1 | 13, 14 | 1536 | ||
NM_001284272.1 | NP_001271201.1 | 13, 14 | 1536 | |||
NM_009445.2 | NP_033471.2 | 13, 14 | 1536 | |||
GenBank mRNA | AK041487.1 | - | 13, 14 | 1507 | ||
AK049200.1 | - | 13, 14 | 1531 | |||
AK164093.1 | - | 13, 14 | 1535 | |||
AX775731.1 | - | 13, 14 | 1502 | |||
BC058851.1 | - | 13, 14 | 1522 | 30023533 | ||
M86377.1 | - | 14, 15 | 1562 |