Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293728.1 | NP_001280657.1 | 11 | 1043 | ||
NM_011656.3 | NP_035786.1 | 12 | 1118 | |||
GenBank mRNA | AF047704.1 | - | 12 | 1052 | ||
AK132488.1 | - | 12 | 1050 | |||
AK171705.1 | - | 11 | 1008 | |||
BC019213.1 | - | 12 | 1049 | 5064658 |