Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122754.2 | NP_001116226.1 | 8 | 731 | ||
NM_001289494.1 | NP_001276423.1 | 7 | 665 | |||
NM_001289495.1 | NP_001276424.1 | 8 | 748 | |||
NM_013698.3 | NP_038726.2 | 7 | 815 | |||
GenBank mRNA | AK088209.1 | - | 7 | 661 | ||
AK138069.1 | - | 8 | 728 | |||
AK138786.1 | - | 8 | 745 | |||
BC120552.1 | - | 7 | 718 | 40129475 | ||
BC137710.1 | - | 7 | 720 | 8860729 | ||
D43963.1 | - | 7 | 815 | |||
D83201.1 | - | 7 | 755 | |||
D83202.1 | - | 7 | 918 | |||
U16145.1 | - | 8 | 696 | |||
U19607.1 | - | 8 | 731 |