Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282014.1 | NP_001268943.1 | 4 | 1171 | ||
NM_001282015.1 | NP_001268944.1 | 4 | 1165 | |||
NM_031202.3 | NP_112479.1 | 4 | 1186 | |||
GenBank mRNA | AK148332.1 | - | 4 | 1171 | ||
AK148370.1 | - | 4 | 1077 | |||
AK148441.1 | - | 4 | 1077 | |||
AK180622.1 | - | 1 | 88 | |||
AK190391.1 | - | 1 | 88 | |||
AK190783.1 | - | 1 | 88 | |||
AK191129.1 | - | 1 | 88 | |||
AK193049.1 | - | 1 | 88 | |||
AK193925.1 | - | 1 | 88 | |||
AK194119.1 | - | 88 | ||||
AK194181.1 | - | 1 | 88 | |||
AK194480.1 | - | 1 | 88 | |||
AK194755.1 | - | 1 | 62 | |||
AK196212.1 | - | 1 | 88 | |||
AK196465.1 | - | 1 | 88 | |||
AK197240.1 | - | 1 | 88 | |||
AK197400.1 | - | 1 | 88 | |||
AK197957.1 | - | 1 | 88 | |||
AK199510.1 | - | 1 | 88 | |||
AK199962.1 | - | 1 | 88 | |||
AK200845.1 | - | 1 | 88 | |||
AK201045.1 | - | 1 | 88 | |||
AK201297.1 | - | 1 | 88 | |||
AK203202.1 | - | 1 | 88 | |||
AK203316.1 | - | 88 | ||||
AK204783.1 | - | 1 | 88 | |||
AK205230.1 | - | 1 | 88 | |||
AK211654.1 | - | 1 | 88 | |||
AK214877.1 | - | 1 | 88 | |||
BC076598.1 | - | 4 | 1169 | 30613975 | ||
X03687.1 | - | 4 | 1071 |