Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205231.1 | NP_001192160.1 | 11 | 1278 | ||
NM_133671.3 | NP_598432.2 | 11 | 1266 | |||
GenBank mRNA | AK078496.1 | - | 11 | 1232 | ||
BC007487.1 | - | 7 | 1500 | 3593924 | ||
BC043071.1 | - | 11 | 1266 | 6403830 | ||
BC094451.1 | - | 9 | 919 | |||
BC106134.1 | - | 10 | 982 | 6309073 | ||
X64587.1 | - | 11 | 1184 |