Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136085.2 | NP_001129557.1 | 15 | 1841 | ||
NM_001276316.1 | NP_001263245.1 | 16 | 1933 | |||
NM_001276317.1 | NP_001263246.1 | 15 | 1836 | |||
NM_009457.4 | NP_033483.2 | 15 | 1836 | |||
GenBank mRNA | AK081808.1 | - | 15 | 1808 | ||
AK088057.1 | - | 15 | 1718 | |||
AK088528.1 | - | 15 | 1718 | |||
AK143416.1 | - | 15 | 1742 | |||
AK171667.1 | - | 15 | 1807 | |||
AK181359.1 | - | 1 | 16 | |||
AK205612.1 | - | 1 | 16 | |||
BC058630.1 | - | 15 | 1782 | 6402368 | ||
BC138200.1 | - | 15 | 1699 | 8861219 | ||
BC145984.1 | - | 15 | 1700 | 40131287 | ||
D10576.1 | - | 15 | 1702 |