Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009469.3 | NP_033495.2 | 21 | 2461 | ||
GenBank mRNA | AF053756.1 | - | 21 | 2153 | ||
AF072370.1 | - | 21 | 2144 | |||
AK122357.1 | - | 20 | 1963 | |||
AK148317.1 | - | 21 | 2454 | |||
AK154939.1 | - | 21 | 2478 | |||
AK156534.1 | - | 21 | 2438 | |||
BC053062.1 | - | 21 | 2121 | |||
BC057121.1 | - | 21 | 2461 | 6834534 | ||
BC059835.1 | - | 21 | 2430 | 6406755 |