Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278605.1 | NP_001265534.1 | 6 | 1449 | ||
NM_009470.5 | NP_033496.1 | 6 | 1505 | |||
GenBank mRNA | AK085460.1 | - | 6 | 1499 | ||
AK144065.1 | - | 6 | 1421 | |||
AK164688.1 | - | 6 | 1421 | |||
AK165507.1 | - | 6 | 1365 | |||
BC012973.1 | - | 5 | 1343 | 4210940 | ||
L33406.1 | - | 6 | 1435 | |||
S79893.1 | - | 1 | 82 |