Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 21 | 2760 | ||
GenBank mRNA | AK083423.1 | - | 10 | 1166 | ||
AK143582.1 | - | 21 | 2695 | |||
AK149964.1 | - | 21 | 2718 | |||
AK163201.1 | - | 4 | 406 | |||
AK164245.1 | - | 12 | 1535 | |||
AK169372.1 | - | 22 | 2739 | |||
AK169933.1 | - | 21 | 2730 | |||
AK171271.1 | - | 21 | 2730 | |||
AK171633.1 | - | 21 | 2713 | |||
AK216305.1 | - | 3 | 174 | |||
BC011341.1 | - | 20 | 2537 | 3661165 | ||
BC066180.1 | - | 21 | 2670 | |||
BC066865.1 | - | 21 | 2670 | |||
L00681.1 | - | 21 | 2723 |