Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 8 | 938 | ||
GenBank mRNA | AK143582.1 | - | 8 | 873 | ||
AK149964.1 | - | 8 | 896 | |||
AK169372.1 | - | 8 | 917 | |||
AK169933.1 | - | 8 | 908 | |||
AK171271.1 | - | 8 | 908 | |||
AK171633.1 | - | 8 | 891 | |||
AK187607.1 | - | 2 | 35 | |||
AK191870.1 | - | 1 | 30 | |||
AK194262.1 | - | 2 | 35 | |||
AK209724.1 | - | 2 | 35 | |||
AK211982.1 | - | 1 | 30 | |||
AK218855.1 | - | 2 | 35 | |||
AK220512.1 | - | 8 | 895 | |||
BC011341.1 | - | 7 | 715 | 3661165 | ||
BC066180.1 | - | 8 | 848 | |||
BC066865.1 | - | 8 | 848 | |||
L00681.1 | - | 8 | 901 |