Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142437.1 | - | 4 | 342 | ||
BC049784.1 | - | 4 | 285 | 6506829 | ||
CJ050979.1 | - | 4 | 421 |