Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252635.1 | NP_001239564.1 | 61 | 9235 | ||
NM_023370.3 | NP_075859.2 | 62 | 9241 | |||
GenBank mRNA | AF308939.1 | - | 60 | 8834 | ||
AJ888000.1 | - | 59 | 8828 | |||
AJ888001.1 | - | 59 | 8828 | |||
AJ888002.1 | - | 59 | 8828 | |||
AK039126.1 | - | 17 | 2576 | |||
AY026062.1 | - | 60 | 8843 | |||
AY563163.1 | - | 14 | 2456 | |||
AY563164.1 | - | 14 | 2456 |