Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009502.4 | NP_033528.3 | 16 | 2267 | ||
GenBank mRNA | AK035184.1 | - | 3 | 298 | ||
AK077850.1 | - | 16 | 2303 | |||
BC008520.1 | - | 16 | 2267 | 3495710 | ||
BC008554.1 | - | 2 | 148 | |||
L18880.1 | - | 16 | 2196 |